Product Details
- SNP ID
-
rs1055812
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:103089869 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAGAGGAGACTCCATCTCATTGA[A/T]ATCTACGTGTGTGTGCTCCACCGTT
- Phenotype
-
MIM: 607803
MIM: 600417
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CNNM2
PubMed Links
Gene Details
- Gene
- CNNM2
- Gene Name
- cyclin and CBS domain divalent metal cation transport mediator 2
There are no transcripts associated with this gene.
- Gene
- NT5C2
- Gene Name
- 5'-nucleotidase, cytosolic II
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134373.2 |
1617 |
Missense Mutation |
ATC,TTC |
I497F |
NP_001127845.1 |
NM_012229.4 |
1617 |
Missense Mutation |
ATC,TTC |
I497F |
NP_036361.1 |
XM_005269632.4 |
1617 |
Missense Mutation |
ATC,TTC |
I505F |
XP_005269689.1 |
XM_005269633.4 |
1617 |
Missense Mutation |
ATC,TTC |
I505F |
XP_005269690.1 |
XM_005269634.4 |
1617 |
Missense Mutation |
ATC,TTC |
I505F |
XP_005269691.1 |
XM_005269635.4 |
1617 |
Missense Mutation |
ATC,TTC |
I505F |
XP_005269692.1 |
XM_005269636.4 |
1617 |
Missense Mutation |
ATC,TTC |
I497F |
XP_005269693.1 |
XM_005269637.4 |
1617 |
Missense Mutation |
ATC,TTC |
I476F |
XP_005269694.1 |
XM_005269638.4 |
1617 |
Missense Mutation |
ATC,TTC |
I473F |
XP_005269695.1 |
XM_005269639.4 |
1617 |
Missense Mutation |
ATC,TTC |
I468F |
XP_005269696.1 |
XM_005269640.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269697.1 |
XM_005269641.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269698.1 |
XM_005269642.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269699.1 |
XM_005269643.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269700.1 |
XM_005269644.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269701.1 |
XM_005269645.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269702.1 |
XM_005269646.4 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_005269703.1 |
XM_006717721.3 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_006717784.1 |
XM_006717723.3 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_006717786.1 |
XM_011539537.2 |
1617 |
Missense Mutation |
ATC,TTC |
I448F |
XP_011537839.1 |
XM_017015947.1 |
1617 |
Missense Mutation |
ATC,TTC |
I474F |
XP_016871436.1 |
XM_017015948.1 |
1617 |
Missense Mutation |
ATC,TTC |
I466F |
XP_016871437.1 |
XM_017015949.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871438.1 |
XM_017015950.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871439.1 |
XM_017015951.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871440.1 |
XM_017015952.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871441.1 |
XM_017015953.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871442.1 |
XM_017015954.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871443.1 |
XM_017015955.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871444.1 |
XM_017015956.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871445.1 |
XM_017015957.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871446.1 |
XM_017015958.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871447.1 |
XM_017015959.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871448.1 |
XM_017015960.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871449.1 |
XM_017015961.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871450.1 |
XM_017015962.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871451.1 |
XM_017015963.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871452.1 |
XM_017015964.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871453.1 |
XM_017015965.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871454.1 |
XM_017015966.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871455.1 |
XM_017015967.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871456.1 |
XM_017015968.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871457.1 |
XM_017015969.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871458.1 |
XM_017015970.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871459.1 |
XM_017015971.1 |
1617 |
Missense Mutation |
ATC,TTC |
I306F |
XP_016871460.1 |
XM_017015972.1 |
1617 |
Missense Mutation |
ATC,TTC |
I259F |
XP_016871461.1 |
XM_017015973.1 |
1617 |
Missense Mutation |
ATC,TTC |
I259F |
XP_016871462.1 |
XM_017015974.1 |
1617 |
Missense Mutation |
ATC,TTC |
I259F |
XP_016871463.1 |
XM_017015975.1 |
1617 |
Missense Mutation |
ATC,TTC |
I259F |
XP_016871464.1 |
XM_017015976.1 |
1617 |
Missense Mutation |
ATC,TTC |
I259F |
XP_016871465.1 |
View Full Product Details