Product Details

SNP ID
rs10897077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60517309 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAACATTAAATAGCTTTCTGTTT[A/C]AGTTTATATGAAAATAAAATTTATG
Phenotype
MIM: 606550
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MS4A12 PubMed Links

Gene Details

Gene
MS4A12
Gene Name
membrane spanning 4-domains A12
There are no transcripts associated with this gene.

Gene
MS4A13
Gene Name
membrane spanning 4-domains A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012417.2 Intron NP_001012417.2
NM_001100909.1 Intron NP_001094379.1
NM_001278320.1 Intron NP_001265249.1

View Full Product Details