Product Details

SNP ID
rs12895353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64711197 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGATGGGGTTTGAACTTTTGGA[A/G]CCCGCTCTTAGCCCTACCCCACACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG3 PubMed Links

Gene Details

Gene
PLEKHG3
Gene Name
pleckstrin homology and RhoGEF domain containing G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308147.1 Intron NP_001295076.1
XM_011536624.2 Intron XP_011534926.1
XM_011536625.2 Intron XP_011534927.1
XM_011536627.2 Intron XP_011534929.1
XM_011536628.2 Intron XP_011534930.1
XM_011536629.2 Intron XP_011534931.1
XM_017021154.1 Intron XP_016876643.1
XM_017021155.1 Intron XP_016876644.1
XM_017021156.1 Intron XP_016876645.1
XM_017021157.1 Intron XP_016876646.1
XM_017021158.1 Intron XP_016876647.1

View Full Product Details