Product Details

SNP ID
rs1978416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77761558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATGAGACTAGCTAATGGAGAGT[C/T]CTTGGACTCTCTGAAAACATTCACA
Phenotype
MIM: 603055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf178 PubMed Links

Gene Details

Gene
C14orf178
Gene Name
chromosome 14 open reading frame 178
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173978.1 Intron NP_001167449.1
NM_174943.3 Intron NP_777603.1
Gene
SNW1
Gene Name
SNW domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318844.1 Intron NP_001305773.1
NM_012245.2 Intron NP_036377.1
XM_005267413.3 Intron XP_005267470.1

View Full Product Details