Product Details
- SNP ID
-
rs2857332
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:105919386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTGGGGACTCCACTGTGCTCTGG[C/G]TGCTGGGGGTCGTGTGAGCTGAGCA
- Phenotype
-
MIM: 146910
MIM: 147070
MIM: 147010
MIM: 616623
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IGH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79497195] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IGH
- Gene Name
- immunoglobulin heavy locus
There are no transcripts associated with this gene.
- Gene
- KIAA0125
- Gene Name
- KIAA0125
There are no transcripts associated with this gene.
View Full Product Details