Product Details
- SNP ID
-
rs6605610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:85248622 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGAGAGGAGGAAATTTTTTTTGG[A/G]AAAATTGAAAAAGGGGACCTCTTTA
- Phenotype
-
MIM: 611436
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CA13
PubMed Links
Gene Details
- Gene
- CA13
- Gene Name
- carbonic anhydrase 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198584.2 |
|
Intron |
|
|
NP_940986.1 |
View Full Product Details