Product Details

SNP ID
rs12756816
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:168180752 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGGAAAAAAATTTCCCTTCATC[C/T]CACCAGAAATTGAATTACTTTTTTG
Phenotype
MIM: 611807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIPRL PubMed Links

Gene Details

Gene
TIPRL
Gene Name
TOR signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031800.2 Intron NP_001026970.1
NM_152902.4 Intron NP_690866.1

View Full Product Details