Product Details

SNP ID
rs2914958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93725845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTCTATATAGAGAAATGGTGGG[A/G]GTAGGGTAGAAAAGATGGAGCTAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM92A1 PubMed Links

Gene Details

Gene
FAM92A1
Gene Name
family with sequence similarity 92 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283034.1 11485 Intron NP_001269963.1
NM_145269.4 11485 Intron NP_660312.2
XM_005250787.2 11485 Intron XP_005250844.1
Gene
RBM12B
Gene Name
RNA binding motif protein 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203390.2 11485 Intron NP_976324.2
XM_005250915.4 11485 Intron XP_005250972.1
XM_011517027.2 11485 UTR 3 XP_011515329.1
XM_011517028.2 11485 Intron XP_011515330.1
XM_011517029.2 11485 Intron XP_011515331.1
XM_017013406.1 11485 Intron XP_016868895.1
XM_017013407.1 11485 Intron XP_016868896.1
XM_017013408.1 11485 Intron XP_016868897.1

View Full Product Details