Product Details

SNP ID
rs36117362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93726401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTATCATTACAATAGGTTTTCC[A/G]AAATTCTCTGTATGCACCAGATTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM92A1 PubMed Links

Gene Details

Gene
FAM92A1
Gene Name
family with sequence similarity 92 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283034.1 10929 Intron NP_001269963.1
NM_145269.4 10929 Intron NP_660312.2
XM_005250787.2 10929 Missense Mutation CAA,CGA Q152R XP_005250844.1
Gene
RBM12B
Gene Name
RNA binding motif protein 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203390.2 10929 Intron NP_976324.2
XM_005250915.4 10929 Intron XP_005250972.1
XM_011517027.2 10929 UTR 3 XP_011515329.1
XM_011517028.2 10929 Intron XP_011515330.1
XM_011517029.2 10929 Intron XP_011515331.1
XM_017013406.1 10929 Intron XP_016868895.1
XM_017013407.1 10929 Intron XP_016868896.1
XM_017013408.1 10929 Intron XP_016868897.1

View Full Product Details