Product Details

SNP ID
rs17783705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93728093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTACTAAAAATTATGACATGGAC[A/T]TTTTTTTGAAATATTTGTTGTGTGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM92A1 PubMed Links

Gene Details

Gene
FAM92A1
Gene Name
family with sequence similarity 92 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283034.1 9237 Intron NP_001269963.1
NM_145269.4 9237 Intron NP_660312.2
XM_005250787.2 9237 Intron XP_005250844.1
Gene
RBM12B
Gene Name
RNA binding motif protein 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203390.2 9237 Intron NP_976324.2
XM_005250915.4 9237 Intron XP_005250972.1
XM_011517027.2 9237 UTR 3 XP_011515329.1
XM_011517028.2 9237 Intron XP_011515330.1
XM_011517029.2 9237 Intron XP_011515331.1
XM_017013406.1 9237 Intron XP_016868895.1
XM_017013407.1 9237 Intron XP_016868896.1
XM_017013408.1 9237 Intron XP_016868897.1

View Full Product Details