Product Details

SNP ID
rs4736596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132561166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCTGTTGGTTACCCAAGGCAGC[A/T]ATTGCATGAACAATCTCTAAACAAA
Phenotype
MIM: 614930
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HPYR1 PubMed Links

Gene Details

Gene
HPYR1
Gene Name
Helicobacter pylori responsive 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
LRRC6
Gene Name
leucine rich repeat containing 6
There are no transcripts associated with this gene.

View Full Product Details