Product Details
- SNP ID
-
rs531425
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:9991647 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCAAATCTGCTTAATTATTGATA[A/G]GCTAGAAAAAAGATAAAGATTCAAG
- Phenotype
-
MIM: 612088
MIM: 606783
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC12A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115518238] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLEC12A
- Gene Name
- C-type lectin domain family 12 member A
- Gene
- CLEC1B
- Gene Name
- C-type lectin domain family 1 member B
View Full Product Details