Product Details

SNP ID
rs1117261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85099577 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCAAGTGTGATTTATCCTAGGAA[C/T]GTAAGGGTGCACCAACATAAGAATA
Phenotype
MIM: 300955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOOL PubMed Links

Gene Details

Gene
APOOL
Gene Name
apolipoprotein O like
There are no transcripts associated with this gene.

Gene
SATL1
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012980.2 Intron NP_001012998.2
XM_011530940.2 Intron XP_011529242.1

View Full Product Details