Product Details

SNP ID
rs13047780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46313656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTTCTGTCCTGGGAGGAGGTT[C/T]CTGGTGCTGGAAGACAGGAAGGCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C21orf58 PubMed Links

Gene Details

Gene
C21orf58
Gene Name
chromosome 21 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286462.1 1300 Intron NP_001273391.1
NM_001286463.1 1300 Intron NP_001273392.1
NM_001286476.1 1300 Intron NP_001273405.1
NM_001286477.1 1300 Intron NP_001273406.1
NM_058180.4 1300 Intron NP_478060.2
XM_005261148.4 1300 Intron XP_005261205.1
XM_006724018.3 1300 Intron XP_006724081.1
XM_011529619.2 1300 Intron XP_011527921.1
XM_011529620.2 1300 Intron XP_011527922.1
XM_011529621.2 1300 Intron XP_011527923.1
XM_011529623.2 1300 Intron XP_011527925.1
XM_011529624.2 1300 Intron XP_011527926.1
XM_011529625.2 1300 Intron XP_011527927.1
XM_011529626.2 1300 Intron XP_011527928.1
XM_011529627.2 1300 Intron XP_011527929.1
XM_011529628.1 1300 Intron XP_011527930.1
XM_017028388.1 1300 Intron XP_016883877.1
XM_017028389.1 1300 Intron XP_016883878.1
XM_017028390.1 1300 Intron XP_016883879.1
XM_017028391.1 1300 Intron XP_016883880.1
XM_017028392.1 1300 Intron XP_016883881.1
XM_017028393.1 1300 Intron XP_016883882.1
Gene
YBEY
Gene Name
ybeY metallopeptidase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006114.2 1300 Intron NP_001006114.1
NM_001314022.1 1300 Intron NP_001300951.1
NM_001314023.1 1300 Intron NP_001300952.1
NM_001314024.1 1300 Intron NP_001300953.1
NM_001314025.1 1300 Intron NP_001300954.1
NM_001314026.1 1300 Intron NP_001300955.1
NM_058181.2 1300 Intron NP_478061.1
XM_005261156.3 1300 Intron XP_005261213.2
XM_011529629.2 1300 Intron XP_011527931.1
XM_011529630.2 1300 Intron XP_011527932.1
XM_011529631.2 1300 Intron XP_011527933.1
XM_011529633.2 1300 UTR 3 XP_011527935.1
XM_011529634.2 1300 Intron XP_011527936.2
XM_011529635.2 1300 Intron XP_011527937.1
XM_017028394.1 1300 UTR 3 XP_016883883.1
XM_017028395.1 1300 Intron XP_016883884.1
XM_017028396.1 1300 Intron XP_016883885.1

View Full Product Details