Product Details
- SNP ID
-
rs7571968
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:175927031 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAAGTCCTTTGGTGTTGGGAAGAA[A/C]CCCTAGGGAAAGAATATGCAGAATG
- Phenotype
-
MIM: 610236
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LNPK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116791538] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LNPK
- Gene Name
- lunapark, ER junction formation factor
View Full Product Details