Product Details

SNP ID
rs35783446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:144395491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCCCTCGTGCCAAAATTTTGA[C/T]TGAGATCCTACACTTTTTCCCCCCC
Phenotype
MIM: 605802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZEB2 PubMed Links

Gene Details

Gene
ZEB2
Gene Name
zinc finger E-box binding homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171653.1 Intron NP_001165124.1
NM_014795.3 Intron NP_055610.1
XM_006712881.3 Intron XP_006712944.1
XM_006712882.3 Intron XP_006712945.1
XM_011512232.2 Intron XP_011510534.1
XM_017005414.1 Intron XP_016860903.1
XM_017005415.1 Intron XP_016860904.1

View Full Product Details