Product Details

SNP ID
rs7088341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87668063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACCTGGAAGAAAATCCGAGATT[C/T]TATCCCTATAAACTTTATTCTTCTA
Phenotype
MIM: 603005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAPSS2 PubMed Links

Gene Details

Gene
PAPSS2
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015880.1 Intron NP_001015880.1
NM_004670.3 Intron NP_004661.2

View Full Product Details