Product Details
- SNP ID
-
rs9989450
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:22299650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGATACAGTCTTGAAGAGGAAGCC[A/G]GGAATGGGAGATGGCAACAAGCCCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM27E5
PubMed Links
Gene Details
- Gene
- FAM27E5
- Gene Name
- family with sequence similarity E5
There are no transcripts associated with this gene.
View Full Product Details