Product Details

SNP ID
rs6455609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158870458 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCTGTGACGCTGAACACATCAG[C/G]AAACCTCAAACTTTCCCCATCTTTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf99 PubMed Links

Gene Details

Gene
C6orf99
Gene Name
chromosome 6 open reading frame 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195032.1 Intron NP_001181961.1
NM_001302839.1 Intron NP_001289768.1
NM_001302840.1 Intron NP_001289769.1
NM_001302841.1 Intron NP_001289770.1
XM_005266788.4 Intron XP_005266845.1
XM_011535374.2 Intron XP_011533676.1
XM_017010142.1 Intron XP_016865631.1
XM_017010143.1 Intron XP_016865632.1
XM_017010144.1 Intron XP_016865633.1
XM_017010145.1 Intron XP_016865634.1
XM_017010146.1 Intron XP_016865635.1
XM_017010147.1 Intron XP_016865636.1

View Full Product Details