Product Details

SNP ID
rs3777679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:139149446 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATCGTTGTGCCATGATTTGGGG[A/T]TTTTAGTTTTTTCTAGGTGAGTTTT
Phenotype
MIM: 607977 MIM: 611438
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HECA PubMed Links

Gene Details

Gene
HECA
Gene Name
hdc homolog, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016217.2 Intron NP_057301.1
Gene
TXLNB
Gene Name
taxilin beta
There are no transcripts associated with this gene.

View Full Product Details