Product Details

SNP ID
rs3094298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8382685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCAATTCTCTTAACTATAACCT[C/T]ACGTTTTAGAAGGTACAATCAACGT
Phenotype
MIM: 603704
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRBA2 PubMed Links

Gene Details

Gene
KRBA2
Gene Name
KRAB-A domain containing 2
There are no transcripts associated with this gene.

Gene
RNF222
Gene Name
ring finger protein 222
There are no transcripts associated with this gene.

Gene
RPL26
Gene Name
ribosomal protein L26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000987.3 Intron NP_000978.1
NM_001315530.1 Intron NP_001302459.1
NM_001315531.1 Intron NP_001302460.1

View Full Product Details