Product Details
- SNP ID
-
rs3100898
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:49894043 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCGCATCGATGAGGCTCGCCAGGAG[C/A]TGCACCGCATTATCAATGACCGGGA
- Phenotype
-
MIM: 600464
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARF6
PubMed Links
Gene Details
- Gene
- ARF6
- Gene Name
- ADP ribosylation factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001663.3 |
928 |
Missense Mutation |
ATG,CTG |
M103L |
NP_001654.1 |
View Full Product Details