Product Details

SNP ID
rs3109904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58812942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTACTCAACAATTATTTTGACT[A/C]CTTACCATGATCAAGGCATTGTTAA
Phenotype
MIM: 606863
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TOX PubMed Links

Gene Details

Gene
TOX
Gene Name
thymocyte selection associated high mobility group box
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014729.2 Intron NP_055544.1
XM_017014085.1 Intron XP_016869574.1

View Full Product Details