Product Details

SNP ID
rs3105859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49131311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAATGCAGCTTCTTAATTTTTAT[A/C]TTCTAGAAAGCTACCAGCAACTGTG
Phenotype
MIM: 604508
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COPS2 PubMed Links

Gene Details

Gene
COPS2
Gene Name
COP9 signalosome subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143887.1 Intron NP_001137359.1
NM_004236.3 Intron NP_004227.1

View Full Product Details