Product Details

SNP ID
rs3112996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95146627 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATATACCACACACGGGGGGTGAG[A/G]ATTTATCTTGTAGGCCGTGCAAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF514 PubMed Links

Gene Details

Gene
ZNF514
Gene Name
zinc finger protein 514
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318005.1 4679 UTR 3 NP_001304934.1
NM_032788.2 4679 UTR 3 NP_116177.1
XM_006712806.2 4679 UTR 3 XP_006712869.1

View Full Product Details