Product Details

SNP ID
rs3127157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158843314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGTCACCAGGCAGAGGCATAAC[G/T]GCCTCTACTGCTAGAAGGAATATCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OSTCP1 PubMed Links

Gene Details

Gene
OSTCP1
Gene Name
oligosaccharyltransferase complex subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details