Product Details

SNP ID
rs3129998
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30689982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCAATCCACGGCACCCCTCCC[A/G]CACTTGGTCTCCCTTGGGGACTCAA
Phenotype
MIM: 607593 MIM: 610990
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MDC1 PubMed Links
Additional Information
For this assay, SNP(s) [rs140648860] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MDC1
Gene Name
mediator of DNA damage checkpoint 1
There are no transcripts associated with this gene.

Gene
NRM
Gene Name
nurim (nuclear envelope membrane protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270707.1 Intron NP_001257636.1
NM_001270708.1 Intron NP_001257637.1
NM_001270709.1 Intron NP_001257638.1
NM_001270710.1 Intron NP_001257639.1
NM_007243.2 Intron NP_009174.1
XM_017010224.1 Intron XP_016865713.1
XM_017010225.1 Intron XP_016865714.1
Gene
PPP1R18
Gene Name
protein phosphatase 1 regulatory subunit 18
There are no transcripts associated with this gene.

View Full Product Details