Product Details
- SNP ID
-
rs3177780
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31879762 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCCAACACAAAAATCGCAGTTT[A/T]TTACCAAACCCAACATTTATTGAGA
- Phenotype
-
MIM: 604599
MIM: 606107
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EHMT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112211188] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EHMT2
- Gene Name
- euchromatic histone lysine methyltransferase 2
- Gene
- SLC44A4
- Gene Name
- solute carrier family 44 member 4
View Full Product Details