Product Details

SNP ID
rs3170843
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:52032984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGGCGAGGGGCTGGGACTGCG[C/T]AGACTGATGGGGTGGTGGGGCCCGG
Phenotype
MIM: 609448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KTI12 PubMed Links

Gene Details

Gene
KTI12
Gene Name
KTI12 chromatin associated homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138417.2 817 Missense Mutation ACG,GCG T260A NP_612426.1
Gene
TXNDC12
Gene Name
thioredoxin domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015913.3 817 Intron NP_056997.1

View Full Product Details