Product Details

SNP ID
rs3172959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140558404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGCCAGGGTCCTACGTTGTGG[C/T]GCAGTGCCTCCCAGTCATCCGTATA
Phenotype
MIM: 602711 MIM: 603483 MIM: 603819
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKHD1-EIF4EBP3 PubMed Links

Gene Details

Gene
ANKHD1-EIF4EBP3
Gene Name
ANKHD1-EIF4EBP3 readthrough
There are no transcripts associated with this gene.

Gene
APBB3
Gene Name
amyloid beta precursor protein binding family B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006051.3 1995 UTR 3 NP_006042.3
NM_133172.2 1995 UTR 3 NP_573418.2
NM_133173.2 1995 UTR 3 NP_573419.2
NM_133174.2 1995 UTR 3 NP_573420.2
Gene
EIF4EBP3
Gene Name
eukaryotic translation initiation factor 4E binding protein 3
There are no transcripts associated with this gene.

Gene
MIR6831
Gene Name
microRNA 6831
There are no transcripts associated with this gene.

Gene
SLC35A4
Gene Name
solute carrier family 35 member A4
There are no transcripts associated with this gene.

Gene
SRA1
Gene Name
steroid receptor RNA activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035235.3 1995 Intron NP_001030312.2
NM_001253764.1 1995 Intron NP_001240693.1

View Full Product Details