Product Details

SNP ID
rs3187207
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:107572795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTGAGGGTAAAAGTGTTTGCC[A/G]GGCAATGAGTCACCTTGTTGGACAA
Phenotype
MIM: 605541
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAV3 PubMed Links

Gene Details

Gene
VAV3
Gene Name
vav guanine nucleotide exchange factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079874.1 2010 UTR 3 NP_001073343.1
NM_006113.4 2010 UTR 3 NP_006104.4
XM_005270360.2 2010 UTR 3 XP_005270417.1
XM_005270361.1 2010 Intron XP_005270418.1
XM_017000053.1 2010 UTR 3 XP_016855542.1
XM_017000054.1 2010 Intron XP_016855543.1
XM_017000055.1 2010 UTR 3 XP_016855544.1
XM_017000056.1 2010 UTR 3 XP_016855545.1

View Full Product Details