Product Details

SNP ID
rs3180761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99395742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCTGCTTCAGCCTACTGTGGC[C/T]TGGAAGGGTCCCTGCCTGCTGAGGC
Phenotype
MIM: 604223 MIM: 607075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC1B PubMed Links

Gene Details

Gene
ARPC1B
Gene Name
actin related protein 2/3 complex subunit 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005720.3 1620 Intron NP_005711.1
XM_006715825.1 1620 Intron XP_006715888.1
XM_006715826.1 1620 Intron XP_006715889.1
Gene
PDAP1
Gene Name
PDGFA associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014891.6 1620 UTR 3 NP_055706.1

View Full Product Details