Product Details

SNP ID
rs3182941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27830697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAACAAACCTCTGTTGCTTTCA[A/G]ACTTAATGTGCTCTGTCCCCAACAA
Phenotype
MIM: 606246 MIM: 605326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JAZF1 PubMed Links

Gene Details

Gene
JAZF1
Gene Name
JAZF zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175061.3 2776 UTR 3 NP_778231.2
XM_006715656.1 2776 UTR 3 XP_006715719.1
XM_017011831.1 2776 UTR 3 XP_016867320.1
XM_017011832.1 2776 UTR 3 XP_016867321.1
Gene
TAX1BP1
Gene Name
Tax1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079864.2 2776 Intron NP_001073333.1
NM_001206901.1 2776 Intron NP_001193830.1
NM_001206902.1 2776 Intron NP_001193831.1
NM_006024.6 2776 Intron NP_006015.4
XM_005249900.2 2776 Intron XP_005249957.1
XM_017012754.1 2776 Intron XP_016868243.1

View Full Product Details