Product Details

SNP ID
rs3176815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34690639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCCCTGATCACCCCAACCTCT[C/T]TCCCAGGACAATCCAGCAAGTAAAG
Phenotype
MIM: 602227
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL19 PubMed Links

Gene Details

Gene
CCL19
Gene Name
C-C motif chemokine ligand 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006274.2 Intron NP_006265.1

View Full Product Details