Product Details
- SNP ID
-
hCV27468494
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:133207298 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTAAAAGATGTAGTGGTCCCTCC[C/T]GGGCAGCTCCTGCAGGTACATGAGC
- Phenotype
-
MIM: 604606
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OBP2B
PubMed Links
Gene Details
- Gene
- OBP2B
- Gene Name
- odorant binding protein 2B
View Full Product Details