Product Details

SNP ID
rs17841107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25119396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATATACATTTATAATTTTCAT[A/T]TGCCACATGGTAAACTGAAAAATAC
Phenotype
MIM: 601491
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IPW PubMed Links

Gene Details

Gene
IPW
Gene Name
imprinted in Prader-Willi syndrome (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details