Product Details

SNP ID
rs3186976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45437223 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACACAGAGGAACCGGGGTCGCCA[C/G]GACTGGCCCTGCCCTGCTGAGGGGC
Phenotype
MIM: 610591 MIM: 611466
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP27 PubMed Links

Gene Details

Gene
ARHGAP27
Gene Name
Rho GTPase activating protein 27
There are no transcripts associated with this gene.

Gene
PLEKHM1
Gene Name
pleckstrin homology and RUN domain containing M1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014798.2 3679 UTR 3 NP_055613.1
XM_006722201.3 3679 UTR 3 XP_006722264.1
XM_011525523.2 3679 Intron XP_011523825.1
XM_011525524.1 3679 UTR 3 XP_011523826.1
XM_011525525.1 3679 UTR 3 XP_011523827.1
XM_011525526.1 3679 UTR 3 XP_011523828.1
XM_011525528.2 3679 UTR 3 XP_011523830.1
XM_017025451.1 3679 UTR 3 XP_016880940.1
XM_017025452.1 3679 UTR 3 XP_016880941.1
XM_017025453.1 3679 UTR 3 XP_016880942.1
XM_017025454.1 3679 UTR 3 XP_016880943.1

View Full Product Details