Product Details

SNP ID
rs3209185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3155680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCGAAGGGTCCAGACCTGACAGA[G/A]TCCATGGTGCCAGGCTCCAGATCCA
Phenotype
MIM: 603156 MIM: 615101
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
BPHL PubMed Links

Gene Details

Gene
BPHL
Gene Name
biphenyl hydrolase-like (serine hydrolase)
There are no transcripts associated with this gene.

Gene
TUBB2A
Gene Name
tubulin beta 2A class IIa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001069.2 308 Silent Mutation GAC,GAT D74D NP_001060.1
NM_001310315.1 308 UTR 5 NP_001297244.1

View Full Product Details