Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022748.11 | 4606 | UTR 3 | NP_073585.8 | ||
XM_011515476.2 | 4606 | UTR 3 | XP_011513778.1 | ||
XM_011515477.2 | 4606 | UTR 3 | XP_011513779.1 | ||
XM_011515478.2 | 4606 | UTR 3 | XP_011513780.1 | ||
XM_011515479.2 | 4606 | UTR 3 | XP_011513781.1 | ||
XM_011515480.2 | 4606 | UTR 3 | XP_011513782.1 | ||
XM_011515481.2 | 4606 | UTR 3 | XP_011513783.1 | ||
XM_011515482.2 | 4606 | UTR 3 | XP_011513784.1 | ||
XM_011515483.2 | 4606 | UTR 3 | XP_011513785.1 | ||
XM_011515484.2 | 4606 | UTR 3 | XP_011513786.1 | ||
XM_017012537.1 | 4606 | UTR 3 | XP_016868026.1 | ||
XM_017012538.1 | 4606 | UTR 3 | XP_016868027.1 | ||
XM_017012539.1 | 4606 | UTR 3 | XP_016868028.1 |