Product Details

SNP ID
rs3734668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13790161 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTGTGCCCTGAACATGGAGACC[C/G]CCAAATCCTCTATGTGCCAAAGATT
Phenotype
MIM: 616952
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MCUR1 PubMed Links

Gene Details

Gene
MCUR1
Gene Name
mitochondrial calcium uniporter regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031713.3 1859 UTR 3 NP_001026883.1
XM_011514802.1 1859 Intron XP_011513104.1

View Full Product Details