Product Details

SNP ID
rs3737511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74148734 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATAGGCTTGAGTACGTGTCCGGC[C/G]GCATGTGTAGTGAACCCTAAAGCTT
Phenotype
MIM: 609093 MIM: 615180
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXO15 PubMed Links

Gene Details

Gene
FBXO15
Gene Name
F-box protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142958.1 208 Intron NP_001136430.1
NM_152676.2 208 Intron NP_689889.1
XM_011525856.1 208 Intron XP_011524158.1
XM_011525858.2 208 Intron XP_011524160.1
XM_011525859.1 208 Intron XP_011524161.1
XM_011525860.1 208 Intron XP_011524162.1
XM_017025603.1 208 Intron XP_016881092.1
XM_017025604.1 208 Intron XP_016881093.1
Gene
TIMM21
Gene Name
translocase of inner mitochondrial membrane 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014177.2 208 UTR 5 NP_054896.2
XM_005266689.3 208 UTR 5 XP_005266746.1

View Full Product Details