Product Details

SNP ID
rs3739029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:134841837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATCATATACTTACACCTTCTTT[C/T]GTTAGCAATTGCACTTCATCGATAT
Phenotype
MIM: 608889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACMSD PubMed Links

Gene Details

Gene
ACMSD
Gene Name
aminocarboxymuconate semialdehyde decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307983.1 Intron NP_001294912.1
NM_138326.2 Intron NP_612199.2
XM_005263586.4 Intron XP_005263643.1
XM_005263588.4 Intron XP_005263645.1
XM_005263589.4 Intron XP_005263646.1
XM_005263590.4 Intron XP_005263647.1
XM_011510592.2 Intron XP_011508894.1
XM_017003325.1 Intron XP_016858814.1
XM_017003326.1 Intron XP_016858815.1

View Full Product Details