Product Details

SNP ID
rs3742818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73716187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCCAGCTTCTATAGCTGCACCTG[C/G]TTTATTGGCAGGTCACTTGCCACAA
Phenotype
MIM: 604010
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ELMSAN1 PubMed Links

Gene Details

Gene
ELMSAN1
Gene Name
ELM2 and Myb/SANT domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043318.2 6656 UTR 3 NP_001036783.1
NM_194278.3 6656 UTR 3 NP_919254.2
XM_005268204.1 6656 UTR 3 XP_005268261.1
XM_005268205.1 6656 UTR 3 XP_005268262.1
XM_005268206.1 6656 UTR 3 XP_005268263.1
Gene
PNMA1
Gene Name
paraneoplastic Ma antigen 1
There are no transcripts associated with this gene.

View Full Product Details