Product Details
- SNP ID
-
rs3744776
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48577068 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAGAGAAAGTTTTATTGCCCCC[A/G]AAAGAGAGAGTCCTTTCTTCCAGGG
- Phenotype
-
MIM: 142966
MIM: 142965
MIM: 610173
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HOXB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79154089] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HOXB3
- Gene Name
- homeobox B3
- Gene
- HOXB4
- Gene Name
- homeobox B4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024015.4 |
|
Intron |
|
|
NP_076920.1 |
- Gene
- MIR10A
- Gene Name
- microRNA 10a
There are no transcripts associated with this gene.
View Full Product Details