Product Details
- SNP ID
-
rs3213641
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:25226804 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGGCAGGTTAGGAACACGTCTCA[C/T]GAGGCTCACCTCCATATTTCTTCCA
- Phenotype
-
MIM: 616984
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPVF
PubMed Links
Gene Details
- Gene
- NPVF
- Gene Name
- neuropeptide VF precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022150.3 |
408 |
Missense Mutation |
ATG,GTG |
M121V |
NP_071433.3 |
View Full Product Details