Product Details

SNP ID
rs3753391
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:160344089 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TAATGTACTTAAGTGTTTGCTGATA[C/T]GCGTTCATTTACTGATACATGGCTT
Phenotype
MIM: 601924 MIM: 605254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 Intron NP_001091868.1
NM_004371.3 Intron NP_004362.2
Gene
NCSTN
Gene Name
nicastrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290184.1 Intron NP_001277113.1
NM_001290186.1 Intron NP_001277115.1
NM_015331.2 Intron NP_056146.1
XM_005245053.4 Intron XP_005245110.1
XM_011509363.2 Intron XP_011507665.1

View Full Product Details