Product Details

SNP ID
rs3757540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122322504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTCCACAAAGGTTTGGTTGGC[C/T]CTTGCTTAAGACAAGTGCCTGCCAC
Phenotype
MIM: 609978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADPS2 PubMed Links

Gene Details

Gene
CADPS2
Gene Name
calcium dependent secretion activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009571.3 Intron NP_001009571.2
NM_001167940.1 Intron NP_001161412.1
NM_017954.10 Intron NP_060424.9
XM_005250695.4 Intron XP_005250752.1
XM_005250696.4 Intron XP_005250753.1
XM_005250697.4 Intron XP_005250754.1
XM_005250698.4 Intron XP_005250755.1
XM_005250699.4 Intron XP_005250756.1
XM_005250700.4 Intron XP_005250757.1
XM_005250701.4 Intron XP_005250758.1
XM_005250702.4 Intron XP_005250759.1
XM_005250703.4 Intron XP_005250760.1
XM_005250704.4 Intron XP_005250761.1
XM_005250705.4 Intron XP_005250762.1
XM_005250706.4 Intron XP_005250763.1
XM_005250707.4 Intron XP_005250764.1
XM_005250708.3 Intron XP_005250765.1
XM_011516695.2 Intron XP_011514997.1
XM_011516696.2 Intron XP_011514998.1
XM_011516697.2 Intron XP_011514999.1
XM_017012794.1 Intron XP_016868283.1
XM_017012795.1 Intron XP_016868284.1
XM_017012796.1 Intron XP_016868285.1
XM_017012797.1 Intron XP_016868286.1
XM_017012798.1 Intron XP_016868287.1
XM_017012799.1 Intron XP_016868288.1
XM_017012800.1 Intron XP_016868289.1

View Full Product Details