Product Details

SNP ID
rs3760330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59707915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGGATGTCTCACCCTGGATGT[A/C]TCACGCTGGCGGTAGCCAGGTTATT
Phenotype
MIM: 608625 MIM: 611753
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTRH2 PubMed Links

Gene Details

Gene
PTRH2
Gene Name
peptidyl-tRNA hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015509.2 Intron NP_001015509.1
NM_016077.4 Intron NP_057161.1
XM_011524887.1 Intron XP_011523189.1
Gene
VMP1
Gene Name
vacuole membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details