Product Details

SNP ID
rs3761728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48988868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTTTACATTTCCATTGTTTTT[G/T]CCAGACTAGAAAGACCTAAAAATCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CWH43 PubMed Links

Gene Details

Gene
CWH43
Gene Name
cell wall biogenesis 43 C-terminal homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286791.1 Intron NP_001273720.1
NM_025087.2 Intron NP_079363.2
XM_011513755.1 Intron XP_011512057.1
XM_011513756.2 Intron XP_011512058.1
XM_011513757.2 Intron XP_011512059.1
XM_011513758.1 Intron XP_011512060.1
XM_011513759.1 Intron XP_011512061.1

View Full Product Details