Product Details

SNP ID
rs3765092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21531329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGATAGTTTTATTCTTTCAGAAA[C/T]AAAATATATATAGGATGCTTAGCTG
Phenotype
MIM: 138571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GYS2 PubMed Links

Gene Details

Gene
GYS2
Gene Name
glycogen synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021957.3 701 Intron NP_068776.2
Gene
SPX
Gene Name
spexin hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030572.3 701 UTR 3 NP_085049.1

View Full Product Details